1,3,5-Tribromobenzene 98%

Warning

AC16108-1000 AC16108-0250
200008-438EA 62.6 USD
200008-438 200008-440
1,3,5-Tribromobenzene 98%
1,3,5-Tribromobenzene
Formula: C₆H₃Br₃
MW: 314.8 g/mol
Boiling Pt: 271 °C (1013 hPa)
Melting Pt: 118…123 °C
Storage Temperature: Ambient
MDL Number: MFCD00000080
CAS Number: 626-39-1
EINECS: 210-947-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR