3-Methoxyphenol 97%

Danger

Synonyms: m-Methoxyphenol1-Hydroxy-3-methoxybenzene

AC16106-5000 AC16106-0250 AC16106-1000 AC16106-0050
200010-004EA 91.4 USD
200010-004 200017-128 103526-050 200017-130
3-Methoxyphenol 97%
3-Methoxyphenol
Formula: C₇H₈O₂
MW: 124.14 g/mol
Boiling Pt: 244 °C (760 mmHg)
Melting Pt: –17 °C
Density: 1.145 g/cm³
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00002267
CAS Number: 150-19-6
EINECS: 205-754-6
UN: 2810
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR