Phenyl isothiocyanate 98%

Danger

AC16097-0025 AC16097-1000 AC16097-5000
103526-046EA 578 USD
103526-046 200001-458 200007-332
Phenyl isothiocyanate 98%
Phenyl isothiocyanate
Formula: C₇H₅NS
MW: 135.19 g/mol
Boiling Pt: 221 °C (760 mmHg)
Melting Pt: –21 °C
Density: 1.1288 g/cm³
Flash Pt: 87 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00004798
CAS Number: 103-72-0
EINECS: 203-138-1
UN: 2927
ADR: 6.1,II
Merck Index: 12,07450

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR