2,4-Dichlorobenzoic acid 98%

Warning

AC16090-0010 AC16090-2500 AC16090-0050 AC16090-1000
103510-774EA 149 USD
103510-774 200034-340 200022-176 200016-084
2,4-Dichlorobenzoic acid 98%
2,4-Dichlorobenzoic acid
Formula: C₇H₄Cl₂O₂
MW: 191.01 g/mol
Boiling Pt: >200 °C (760 mmHg)
Melting Pt: 160…164 °C
Storage Temperature: Ambient
MDL Number: MFCD00002414
CAS Number: 50-84-0
EINECS: 200-067-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR