(1S)-(-)-Borneol 97%

Danger

Synonyms: (1S-endo)-1,7,7-Trimethylbicyclo[2.2.1]heptan-2-ol(-)-BorneolL(-)-Borneol

AC16065-1000 AC16065-0250 AC16065-5000 AC16065-0025
200012-916EA 120 USD
200012-916 103530-042 200049-710 103530-044
(1S)-(-)-Borneol 97%
(1S)-(-)-Borneol
Formula: C₁₀H₁₈O
MW: 154.25 g/mol
Boiling Pt: 210 °C (760 mmHg)
Melting Pt: 204…208 °C
Flash Pt: 65 °C
Storage Temperature: Ambient
MDL Number: MFCD00003759
CAS Number: 464-45-9
EINECS: 207-353-1
UN: 1312
ADR: 4.1,III
Merck Index: 13,01328

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR