1-Pentanol 99%, pure

Warning

Synonyms: 1-Pentyl alcoholn-Amyl alcoholAmyl alcohol1-Hydroxy pentaneButyl carbinol

AC16060-0010 AC16060-0025 AC16060-0251 AC16060-0250
200001-442EA 74.5 USD
200001-442 200001-444 103526-038 200008-764
1-Pentanol 99%, pure
1-Pentanol
Formula: H₃C(CH₂)₃CH₂OH
MW: 88.15 g/mol
Boiling Pt: 137…139 °C (760 mmHg)
Melting Pt: –78 °C
Density: 0.811 g/cm³
Flash Pt: 49 °C
Storage Temperature: Ambient
MDL Number: MFCD00002977
CAS Number: 71-41-0
EINECS: 200-752-1
UN: 1105
ADR: 3,III
Merck Index: 12,07257
https://us.vwr.com/stibo/web/std.lang.all/15/15/731515.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR