1,3,5-Trimethoxybenzene 99%

Warning

Synonyms: O,O,O-1,3,5-Trimethylresorcinol

AC16055-2500 AC16055-0500 AC16055-0100
103509-052EA 416 USD
103509-052 200053-676 200008-444
1,3,5-Trimethoxybenzene 99%
1,3,5-Trimethoxybenzene
Formula: C₉H₁₂O₃
MW: 168.19 g/mol
Boiling Pt: 255 °C (760 mmHg)
Melting Pt: 50…53 °C
Flash Pt: 85 °C
Storage Temperature: Ambient
MDL Number: MFCD00008385
CAS Number: 621-23-8
EINECS: 210-673-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR