3-Chlorotoluene 97%

Warning

AC16045-1000 AC16045-2500 AC16045-0025 AC16045-5000
200023-256EA 43.4 USD
200023-256 200047-438 103530-040 200035-338
3-Chlorotoluene 97%
3-Chlorotoluene
Formula: C₇H₇Cl
MW: 126.59 g/mol
Boiling Pt: 160…162 °C (760 mmHg)
Melting Pt: –48 °C
Density: 1.07 g/cm³
Flash Pt: 50 °C
Storage Temperature: Ambient
MDL Number: MFCD00000595
CAS Number: 108-41-8
EINECS: 203-580-5
UN: 2238
ADR: 3,III
Merck Index: 13,02190

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR