2,5-Dimethylbenzoic acid 98%

Warning

AC16033-0500 AC16033-0100
103509-030EA 230 USD
103509-030 200016-166
2,5-Dimethylbenzoic acid 98%
2,5-Dimethylbenzoic acid
Formula: C₉H₁₀O₂
MW: 150.18 g/mol
Boiling Pt: 268 °C (760 mmHg)
Melting Pt: 131…135 °C
Storage Temperature: Ambient
MDL Number: MFCD00002482
CAS Number: 610-72-0
EINECS: 210-235-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR