2,4-Dimethylbenzoic acid 98%

Warning

AC16032-2500 AC16032-0500 AC16032-0100
200028-284EA 151 USD
200028-284 103510-666 200022-202
2,4-Dimethylbenzoic acid 98%
2,4-Dimethylbenzoic acid
Formula: C₉H₁₀O₂
MW: 150.18 g/mol
Boiling Pt: 267 °C (727 mmHg)
Melting Pt: 123…127 °C
Storage Temperature: Ambient
MDL Number: MFCD00002480
CAS Number: 611-01-8
EINECS: 210-246-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR