1,2,3-Trimethoxybenzene 98%

AC16021-0250 AC16021-1000
200045-622EA 102 USD
200045-622 200039-938
1,2,3-Trimethoxybenzene 98%
1,2,3-Trimethoxybenzene
Formula: C₉H₁₂O₃
MW: 168.19 g/mol
Boiling Pt: 241 °C (1013 hPa)
Melting Pt: 42…45 °C
Density: 1.112 g/cm³
Flash Pt: 112 °C
Storage Temperature: Ambient
MDL Number: MFCD00008358
CAS Number: 634-36-6
EINECS: 211-207-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR