5-Methylanthranil acid 97%

Warning

Synonyms: 6-Amino-m-toluic acid2-Amino-5-methylbenzoic acid

AC16020-0010 AC16020-0050 AC16020-0250
200016-314EA 41.81 USD
200016-314 103510-662 200022-420
5-Methylanthranil acid 97%
5-Methylanthranil acid
Formula: C₈H₉NO₂
MW: 151.16 g/mol
Melting Pt: 174…177 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00007909
CAS Number: 2941-78-8
EINECS: 220-932-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR