3,5-Xylidine 97+%

Danger

Synonyms: 5-Amino-m-xylene3,5-Dimethylaniline

AC16019-0050 AC16019-1000 AC16019-5000
200009-732EA 51.8 USD
200009-732 200029-142 200023-092
3,5-Xylidine 97+%
3,5-Xylidine
Formula: C₈H₁₁N
MW: 121.18 g/mol
Boiling Pt: 220 °C (1013 hPa)
Melting Pt: 9.8 °C
Density: 0.97 g/cm³
Flash Pt: 93 °C
Storage Temperature: Ambient
MDL Number: MFCD00007813
CAS Number: 108-69-0
EINECS: 203-607-0
UN: 1711
ADR: 6.1,II
Merck Index: 12,10217

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR