2,4-Di-tert-butylphenol 97%

Warning

Synonyms: 1,3-Di(dimethylethyl)-4-hydroxybenzene

AC16012-0025 AC16012-0251 AC16012-1000 AC16012-5000
200040-662EA 140 USD
200040-662 103530-038 103530-036 200009-004
2,4-Di-tert-butylphenol 97%
2,4-Di-tert-butylphenol
Formula: C₁₄H₂₂O
MW: 206.33 g/mol
Boiling Pt: 264 °C (760 mmHg)
Melting Pt: 54…59 °C
Flash Pt: 115 °C
Storage Temperature: Ambient
MDL Number: MFCD00008828
CAS Number: 96-76-4
EINECS: 202-532-0
UN: 3077
ADR: 9,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR