2,4,6-Tribromophenol 98%

Warning

AC16010-0050 AC16010-1000 AC16010-5000
200040-586EA 25.9 USD
200040-586 200022-144 200034-302
2,4,6-Tribromophenol 98%
2,4,6-Tribromophenol
Formula: C₆H₃Br₃O
MW: 330.8 g/mol
Boiling Pt: 290 °C (1013 hPa)
Melting Pt: 89…93 °C
Storage Temperature: Ambient
MDL Number: MFCD00002150
CAS Number: 118-79-6
EINECS: 204-278-6
UN: 3077
Merck Index: 12,09744

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR