5-Bromosalicylaldehyde 99%

Warning

Synonyms: 5-Bromo-2-hydroxybenzaldehyde

AC15991-0250 AC15991-1000
200010-708EA 49.4 USD
200010-708 200024-062
5-Bromosalicylaldehyde 99%
5-Bromosalicylaldehyde
Formula: C₇H₅BrO₂
MW: 201.02 g/mol
Melting Pt: 103…107 °C
Flash Pt: 110 °C
Storage Temperature: Ambient
MDL Number: MFCD00003330
CAS Number: 1761-61-1
EINECS: 217-167-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR