2-Methyl-3-buten-2-ol 97%

Danger

Synonyms: 3-Hydroxy-3-methyl-1-butene

AC15967-2500 AC15967-0010
200034-896EA 79.2 USD
200034-896 200009-510
2-Methyl-3-buten-2-ol 97%
2-Methyl-3-buten-2-ol
Formula: C₅H₁₀O
MW: 86.13 g/mol
Boiling Pt: 99 °C (1013 hPa)
Melting Pt: –43 °C
Density: 0.82 g/cm³
Flash Pt: 13 °C
Storage Temperature: Ambient
MDL Number: MFCD00004470
CAS Number: 115-18-4
EINECS: 204-068-4
UN: 1987
ADR: 3,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR