Dimethyl malonate 97%

Synonyms: 1,3-Propanedioic acid dimethyl esterMalonic acid dimethyl ester

AC15956-0010 AC15956-2500 AC15956-0025
103509-020EA 204 USD
103509-020 200012-098 200006-326
Dimethyl malonate 97%
Dimethyl malonate
Formula: C₅H₈O₄
MW: 132.12 g/mol
Boiling Pt: 180…181 °C (760 mmHg)
Melting Pt: –62 °C
Density: 1.156 g/cm³
Flash Pt: 90 °C
Storage Temperature: Ambient
MDL Number: MFCD00008460
CAS Number: 108-59-8
EINECS: 203-597-8
Merck Index: 12,06169

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR