1-Iodo-2-nitrobenzene 97%

Warning

AC15945-0250
200021-856EA 77.1 USD
200021-856
1-Iodo-2-nitrobenzene 97%
1-Iodo-2-nitrobenzene
Formula: C₆H₄INO₂
MW: 249.01 g/mol
Boiling Pt: 289 °C (1013 hPa)
Melting Pt: 47…52 °C
Flash Pt: 110 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00007088
CAS Number: 609-73-4
EINECS: 210-200-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR