1,2-Dimethylimidazole 96+%

Danger

Synonyms: 1,2-Dimethyl-1H-imidazole

AC15944-5000 AC15944-1000
200008-428EA 32 USD
200008-428 200027-588
1,2-Dimethylimidazole 96+%
1,2-Dimethylimidazole
Formula: C₅H₈N₂
MW: 96.13 g/mol
Boiling Pt: 204 °C (1013 hPa)
Melting Pt: 37…39 °C
Density: 1.084 g/cm³
Flash Pt: 93 °C
Storage Temperature: Ambient
MDL Number: MFCD00005294
CAS Number: 1739-84-0
EINECS: 217-101-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR