Benzamide 99%

Warning

AC15933-5000 AC15933-1000
200011-310EA 183 USD
200011-310 200011-306
Benzamide 99%
Benzamide
Formula: C₇H₇NO
MW: 121.14 g/mol
Boiling Pt: 228 °C (760 mmHg)
Melting Pt: 125…129 °C
Flash Pt: 180 °C
Storage Temperature: Ambient
MDL Number: MFCD00007968
CAS Number: 55-21-0
EINECS: 200-227-7
Merck Index: 13,01059

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR