Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Chlorobenzoic acid ≥99%

Supplier: ACROS ORGANICS MS

Warning
Synonyms: Chlorodracyclic acidChlorodracylic acidp-Chlorobenzoic acid

AC15925-0010 AC15925-2500 AC15925-0250
200047-820EA 227.8 USD InStock
200047-820 200010-334 200010-332
4-Chlorobenzoic acid ≥99%
4-Chlorobenzoic acid


Formula: C₇H₅ClO₂
MW: 156.57 g/mol
Boiling Pt: 287 °C (1013 hPa)
Melting Pt: 123 °C
Density: 1.535 g/cm³ (20 °C)
Flash Pt: 238 °C
Storage Temperature: Ambient
MDL Number: MFCD00002531
CAS Number: 74-11-3
EINECS: 200-805-9
Merck Index: 13,02144
Order Now
Specification Test Results
Appearance White to yellow Crystalline powder
Infrared spectrum Conforms
Melting point 240°C to 243°C
Water ≤0.1 % (K.F.)
HPLC ≥98.5 %
Titration with NaOH 98.5 to 101.5 %


 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty