Bromocyclohexane 99%

Synonyms: Cyclohexyl bromide

AC15908-0010 AC15908-2500 AC15908-0025 AC15908-0050
200011-836EA 136 USD
200011-836 103509-006 200024-848 200006-158
Bromocyclohexane 99%
Bromocyclohexane
Formula: C₆H₁₁Br
MW: 163.06 g/mol
Boiling Pt: 166…167 °C (760 mmHg)
Density: 1.33 g/cm³
Flash Pt: 62 °C
Storage Temperature: Ambient
MDL Number: MFCD00003819
CAS Number: 108-85-0
EINECS: 203-622-2
Merck Index: 13,02759

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR