Pyrocatechol 99+%

Danger

Synonyms: o-DihydroxybenzeneCatechol1,2-Dihydroxybenzene

AC15898-0050 AC15898-0100 AC15898-1000 AC15898-0025 AC15898-5000
200005-984EA 32.3 USD
200005-984 103526-000 200005-986 200001-416 103526-362
Pyrocatechol 99+%
Pyrocatechol
Formula: C₆H₆O₂
MW: 110.11 g/mol
Boiling Pt: 245 °C (760 mmHg)
Melting Pt: 103…106 °C
Flash Pt: 131 °C
Storage Temperature: Ambient
MDL Number: MFCD00002188
CAS Number: 120-80-9
EINECS: 204-427-5
UN: 2811
ADR: 6.1,III
Merck Index: 12,08183

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR