p-Xylene 99%, pure

Danger

Synonyms: 1,4-Dimethylbenzene

AC15877-0100
103525-984EA 716.54 USD
103525-984
p-Xylene 99%, pure
p-Xylene
Formula: C₆H₄(CH₃)₂
MW: 106.17 g/mol
Boiling Pt: 138 °C (760 mmHg)
Melting Pt: 13 °C
Density: 0.866 g/cm³
Flash Pt: 25 °C
Storage Temperature: Ambient
MDL Number: MFCD00008556
CAS Number: 106-42-3
EINECS: 203-396-5
UN: 1307
ADR: 3,III
Merck Index: 12,10214
https://us.vwr.com/stibo/web/std.lang.all/87/75/768775.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR