di-Sodium succinate hexahydrate 97%

Warning

Synonyms: Sodium succinate dibasic hexahydrateSuccinic acid disodium salt hexahydrateSodium succinate hexahydrateSuccinic acid disodium hexahydrateSodium Succinate, HexahydrateDisodium succinate hexahydrate1,4-Butanedioic acid disodium salt hexahydrate

AC15875-0010 AC15875-5000 AC15875-1000
200026-838EA 84.2 USD
200026-838 200032-866 200020-754
di-Sodium succinate hexahydrate 97%
di-Sodium succinate hexahydrate
Formula: NaOOCCH₂CH₂COONa·6H₂O
MW: 270.14 g/mol
Melting Pt: 120 °C
Storage Temperature: Ambient
MDL Number: MFCD00149117
CAS Number: 6106-21-4
EINECS: 205-778-7
Merck Index: 13,08754

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR