Succinic acid 99%

Danger

Synonyms: 1,4-Butanedioic acid

AC15874-0025 AC15874-2500 AC15874-0010 AC15874-0100 AC15874-5000
103466-510EA 45.5 USD
103466-510 200001-398 200014-576 103466-508 103466-506
Succinic acid 99%
Succinic acid
Formula: HOOCCH₂CH₂COOH
MW: 118.09 g/mol
Boiling Pt: 235 °C (760 mmHg)
Melting Pt: 185…190 °C
Flash Pt: 206 °C
Storage Temperature: Ambient
MDL Number: MFCD00002789
CAS Number: 110-15-6
EINECS: 203-740-4
ADR: 8,III
Merck Index: 12,09038
https://us.vwr.com/stibo/web/std.lang.all/01/38/2300138.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR