trans-Cinnamic acid 98+%

Warning

Synonyms: (2E)-2-Phenyl-2-propenoic acidtrans-Cinnamic acidCinnamic acid (only trans)

AC15857-0025 AC15857-0050 AC15857-5000 AC15857-1000
103508-990EA 332 USD
103508-990 200007-928 200001-386 200001-388
trans-Cinnamic acid 98+%
trans-Cinnamic acid
Formula: C₉H₈O₂
MW: 148.16 g/mol
Boiling Pt: 300 °C (760 mmHg)
Melting Pt: 131…136 °C
Density: 1.248 g/cm³
Flash Pt: >100 °C
Storage Temperature: Ambient
MDL Number: MFCD00004369
CAS Number: 140-10-3
EINECS: 205-398-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR