1,12-Dibromododecane 96%

AC15845-0050 AC15845-1000 AC15845-5000 AC15845-0250
200033-626EA 207 USD
200033-626 103510-770 200008-372 103510-772
1,12-Dibromododecane 96%
1,12-Dibromododecane
Formula: BrH₂C(CH₂)₁₀CH₂Br
MW: 328.13 g/mol
Boiling Pt: 215 °C (15 mmHg)
Melting Pt: 37…42 °C
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00000226
CAS Number: 3344-70-5
EINECS: 222-096-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR