(±)-2-Methyl-1-butanol 98%

Warning

AC15827-0025 AC15827-1000 AC15827-0250 AC15827-0010
200049-166EA 111 USD
200049-166 200025-144 200049-168 200031-100
(±)-2-Methyl-1-butanol 98%
(±)-2-Methyl-1-butanol
Formula: C₅H₁₂O
MW: 88.15 g/mol
Boiling Pt: 129 °C (1013 hPa)
Melting Pt: –70 °C
Density: 0.81 g/cm³
Flash Pt: 43 °C
Storage Temperature: Ambient
MDL Number: MFCD00004743
CAS Number: 137-32-6
EINECS: 205-289-9
UN: 1105
ADR: 3,III
Merck Index: 12,06109

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR