2-Methylaminoethanol 99%

Danger

Synonyms: N-Methyl-2-ethanolaminN-Methyl-2-aminoethanol

AC15826-0025 AC15826-0100 AC15826-5000
200008-834EA 60.6 USD
200008-834 103526-352 103525-950
2-Methylaminoethanol 99%
2-Methylaminoethanol
Formula: C₃H₉NO
MW: 75.11 g/mol
Boiling Pt: 159 °C (760 mmHg)
Melting Pt: –3 °C
Density: 0.93 g/cm³
Flash Pt: 72 °C
Storage Temperature: Ambient
MDL Number: MFCD00002839
CAS Number: 109-83-1
EINECS: 203-710-0
UN: 2735
ADR: 8,II
Merck Index: 12,06096

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR