Bromoform 96% stabilized

Danger

Synonyms: Tribromomethane

AC15820-0010 AC15820-0050 AC15820-2500 AC15820-0025
103525-942EA 1263 USD
103525-942 200057-238 103525-944 200057-240
Bromoform 96% stabilized
Bromoform
Formula: Br₃CH
MW: 252.73 g/mol
Boiling Pt: 150…151 °C (760 mmHg)
Melting Pt: 8 °C
Density: 2.63 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00000128
CAS Number: 75-25-2
EINECS: 200-854-6
UN: 2515
ADR: 6.1,III
Merck Index: 13,01404
https://us.vwr.com/stibo/web/std.lang.all/78/85/717885.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR