3,5-Dimethoxyphenol 97%

Warning

AC15812-0100 AC15812-0500
200009-724EA 96.2 USD
200009-724 103465-154
3,5-Dimethoxyphenol 97%
3,5-Dimethoxyphenol
Formula: C₈H₁₀O₃
MW: 154.17 g/mol
Boiling Pt: 172…175 °C (17 mmHg)
Melting Pt: 40…45 °C
Flash Pt: 78 °C
Storage Temperature: Ambient
MDL Number: MFCD00008388
CAS Number: 500-99-2
EINECS: 207-917-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR