1,2-Diaminocyclohexane cis- and trans-mixture 99%

Danger

Synonyms: 1,2-Diaminocyclohexane1,2-Cyclohexanediamine

AC15810-2500 AC15810-0010
200008-420EA 63.4 USD
200008-420 200015-528
1,2-Diaminocyclohexane cis- and trans-mixture 99%
1,2-Diaminocyclohexane
Formula: C₆H₁₄N₂
MW: 114.19 g/mol
Boiling Pt: 192 °C (1013 hPa)
Melting Pt: 2…15 °C
Density: 0.93 g/cm³
Flash Pt: 75 °C
MDL Number: MFCD00001491
CAS Number: 694-83-7
EINECS: 211-776-7
UN: 2735

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR