p-Toluonitrile 98+%

Warning

Synonyms: 4-Methylbenzonitrilep-Methylbenzonitrile

AC15802-0050 AC15802-0010 AC15802-1000 AC15802-2500
200038-672EA 108 USD
200038-672 200020-534 200044-586 200044-584
p-Toluonitrile 98+%
p-Toluonitrile
Formula: C₈H₇N
MW: 117.15 g/mol
Boiling Pt: 103…106 °C (20 mmHg)
Melting Pt: 26…28 °C
Density: 0.98 g/cm³
Flash Pt: 85 °C
MDL Number: MFCD00001827
CAS Number: 104-85-8
EINECS: 203-244-8
Merck Index: 12,09676

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR