Pyrimidine 99%

Warning

AC15770-0250 AC15770-0050
200020-560EA 100 USD
200020-560 103531-466
Pyrimidine 99%
Pyrimidine
Formula: C₄H₄N₂
Boiling Pt: 123…124 °C (760 mmHg)
Melting Pt: 20…22 °C
Density: 1.01 g/cm³
Flash Pt: 31 °C
Storage Temperature: Ambient
MDL Number: MFCD00006059
CAS Number: 289-95-2
EINECS: 206-026-0
UN: 1993
ADR: 3,III
Merck Index: 12,08170

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR