Fumaronitrile 98%

Danger

AC15740-1000 AC15740-0250 AC15740-0050
200037-490EA 241 USD
200037-490 200025-434 103531-456
Fumaronitrile 98%
Fumaronitrile
Formula: C₄H₂N₂
MW: 78.07 g/mol
Boiling Pt: 186 °C (760 mmHg)
Melting Pt: 95…98 °C
Storage Temperature: Ambient
MDL Number: MFCD00001928
CAS Number: 764-42-1
EINECS: 212-122-3
UN: 3439
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR