5-Aminoindan 97%

Warning

Synonyms: 5-Aminoindane

AC15735-0500 AC15735-0100
200010-678EA 90.6 USD
200010-678 200024-038
5-Aminoindan 97%
5-Aminoindan
Formula: C₉H₁₁N
MW: 133.19 g/mol
Boiling Pt: 247…249 °C (745 mmHg)
Melting Pt: 34…36 °C
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00003803
CAS Number: 24425-40-9
EINECS: 246-241-7
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR