3-Butyne-1-ol 97%

Warning

Synonyms: 4-Hydroxy-1-butyne3-Butyn-1-olBut-3-yn-1-ol1-Hydroxy-3-butyne

AC15733-1000 AC15733-0250 AC15733-0050
200005-234EA 42.7 USD
200005-234 200001-342 200035-290
3-Butyne-1-ol 97%
3-Butyne-1-ol
Formula: C₄H₆O
MW: 70.09 g/mol
Boiling Pt: 128.8 °C (1013 hPa)
Melting Pt: –63.6 °C
Density: 0.926 g/cm³
Flash Pt: 36 °C
Storage Temperature: Ambient
MDL Number: MFCD00002955
CAS Number: 927-74-2
EINECS: 213-161-9
UN: 1986
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR