1-Adamantanol 99%

Synonyms: Adamantan-1-ol1-Hydroxytricyclo[3.3.1.1(3,7)]decaneTricyclo[3.3.1.1(3,7)]decan-1-ol1-Tricyclo[3.3.1.1(3,7)]decanol

AC15710-0050 AC15710-0250 AC15710-1000
103510-756EA 221 USD
103510-756 200008-556 200015-680
1-Adamantanol 99%
1-Adamantanol
Formula: C₁₀H₁₆O
MW: 152.24 g/mol
Melting Pt: >240 °C (subl.)
Storage Temperature: Ambient
MDL Number: MFCD00074729
CAS Number: 768-95-6
EINECS: 212-202-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR