1-Methylpiperazine 99%

Danger

AC15691-5000 AC15691-0050 AC15691-1000 AC15691-0025
200004-776EA 37 USD
200004-776 200004-774 200001-326 103525-900
1-Methylpiperazine 99%
1-Methylpiperazine
Formula: C₅H₁₂N₂
MW: 100.16 g/mol
Boiling Pt: 138 °C (760 mmHg)
Melting Pt: –6 °C
Density: 0.903 g/cm³
Flash Pt: 42 °C
Storage Temperature: Ambient
MDL Number: MFCD00005966
CAS Number: 109-01-3
EINECS: 203-639-5
UN: 2734
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR