m-Anisaldehyde 97%

Warning

Synonyms: 3-Methoxybenzaldehyde

AC15676-0250 AC15676-1000 AC15676-5000
200013-184EA 135 USD
200013-184 200006-930 103510-748
m-Anisaldehyde 97%
m-Anisaldehyde
Formula: C₈H₈O₂
MW: 136.15 g/mol
Boiling Pt: 143 °C (50 mmHg)
Density: 1.119 g/cm³
Flash Pt: 110 °C
Storage Temperature: Ambient
MDL Number: MFCD00003361
CAS Number: 591-31-1
EINECS: 209-712-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR