4-Iodoaniline 99%

Warning

AC15661-1000 AC15661-5000 AC15661-0250
200005-464EA 53 USD
200005-464 103531-448 200005-462
4-Iodoaniline 99%
4-Iodoaniline
Formula: C₆H₆IN
MW: 219.02 g/mol
Boiling Pt: 262 °C (1013 hPa)
Melting Pt: 59…64 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00007848
CAS Number: 540-37-4
EINECS: 208-743-4
UN: 2811
ADR: 6.1,III
Merck Index: 13,05049

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR