Tribenzylamine 99+%

Warning

AC15647-0010 AC15647-2500 AC15647-1000
200021-020EA 53 USD
200021-020 200014-930 103510-738
Tribenzylamine 99+%
Tribenzylamine
Formula: N(CH₂C₆H₅)₃
MW: 287.4 g/mol
Boiling Pt: 380…390 °C (760 mmHg)
Melting Pt: 90…94 °C
Storage Temperature: Ambient
MDL Number: MFCD00004773
CAS Number: 620-40-6
EINECS: 210-638-3
ADR: 4.1,III
https://us.vwr.com/stibo/web/std.lang.all/70/36/767036.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR