6-Methyl-2-pyridone 97%

Synonyms: 6-Methylpyridin-2-ol2-Hydroxy-6-methylpyridine6-Methylpyridin-2(1H)-one

AC15639-0050 AC15639-0250
200022-704EA 159 USD
200022-704 200046-912
6-Methyl-2-pyridone 97%
6-Methyl-2-pyridone
Formula: C₆H₇NO
MW: 109.13 g/mol
Boiling Pt: 283…284 °C (1013 hPa)
Melting Pt: 155…160 °C
MDL Number: MFCD00006279
CAS Number: 3279-76-3
EINECS: 221-919-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR