4-Hydroxy-3-methoxycinnamic acid (Ferulic acid) 99%

Synonyms: NSC 51986Coniferic acidNSC 2821NSC 6743203-(4-Hydroxy-3-methoxyphenyl)acrylic acidFerulic acid

AC15636-1000 AC15636-0050 AC15636-0250
200017-576EA 74.6 USD
200017-576 200035-918 200010-454
4-Hydroxy-3-methoxycinnamic acid (Ferulic acid) 99%
4-Hydroxy-3-methoxycinnamic acid (Ferulic acid)
Formula: C₁₀H₁₀O₄
MW: 194.18 g/mol
CAS Number: 1135-24-6
EINECS: 208-679-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR