2-Hydroxyethyl methacrylate ≥97% stabilized

Supplier: ACROS ORGANICS MS

Warning

Synonyms: HEMAMethacrylic acid 2-hydroxyethyl esterGMAGlycol methacrylate1,2-Ethanediol mono(2-methylpropenoate)

AC15633-0025 AC15633-0010 AC15633-2500 AC15633-5000
103510-734EA 156.11 USD
103510-734 200056-882 103510-736 200056-890
2-Hydroxyethyl methacrylate ≥97% stabilized
2-Hydroxyethyl methacrylate
Formula: C₆H₁₀O₃
MW: 130.14 g/mol
Boiling Pt: 205…208 °C (760 mmHg)
Melting Pt: –12 °C
Density: 1,073
Flash Pt: 101 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00002863
CAS Number: 868-77-9
EINECS: 212-782-2

Order Now

Specification Test Results

Appearance Clear liquid
Infrared spectrum Conforms
Refractive index 1.4510 to 1.4530 (20°C, 589 nm)
Water ≤0.2 % (K.F.)
GC ≥96.0 %
Color scale ≤15 APHA
Stabilizer 200 to 400 ppm MEHQ
Acid value ≤2 mg KOH/g

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR