4-Fluorobenzaldehyde 98+%

Warning

AC15615-0010 AC15615-2500 AC15615-0500
200005-432EA 82.6 USD
200005-432 200010-412 103525-870
4-Fluorobenzaldehyde 98+%
4-Fluorobenzaldehyde
Formula: C₇H₅FO
MW: 124.11 g/mol
Boiling Pt: 181 °C (758 mmHg)
Melting Pt: –10 °C
Density: 1.15 g/cm³
Flash Pt: 56 °C
Storage Temperature: Ambient
MDL Number: MFCD00003378
CAS Number: 459-57-4
EINECS: 207-293-6
UN: 1989
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR