2-Ethoxyethanol 99%, Extra Pure

Danger

Synonyms: CellosolveEthylene glycol monoethyl ether

AC15602-0025 AC15602-0250 AC15602-0010
103525-866EA 806.54 USD
103525-866 200001-312 103529-090
2-Ethoxyethanol 99%, Extra Pure
2-Ethoxyethanol
Formula: C₂H₅OCH₂CH₂OH
MW: 90.12 g/mol
Boiling Pt: 135 °C (760 mmHg)
Melting Pt: –70 °C
Density: 0.93 g/cm³
Flash Pt: 40 °C
Storage Temperature: Ambient
MDL Number: MFCD00002869
CAS Number: 110-80-5
EINECS: 203-804-1
UN: 1171
ADR: 3,III
REACH: 01-2119560582-38
Merck Index: 13,03786
https://us.vwr.com/stibo/web/std.lang.all/93/02/2299302.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR