4-Methylbenzyl alcohol 98%

Warning

Synonyms: p-Tolylmethanol4-(Hydroxymethyl)toluole(4-Methylphenyl)methanol

AC15592-1000 AC15592-0250 AC15592-0050
200010-546EA 48.5 USD
200010-546 200047-990 103510-638
4-Methylbenzyl alcohol 98%
4-Methylbenzyl alcohol
Formula: C₈H₁₀O
MW: 122.17 g/mol
Boiling Pt: 217 °C (760 mmHg)
Melting Pt: 56…61 °C
Storage Temperature: Ambient
MDL Number: MFCD00004664
CAS Number: 589-18-4
EINECS: 209-639-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR